Would you like to log in again?
Avantor® Hichrom HI-DEX DET Beta, GC Columns
Supplier: VWR
Avantor® Hichrom
HI80-10-010-5 HI68-25-015-60 HI68-25-015-25 HI68-25-025-60 HI68-25-015-30 HI68-25-025-30 HI68-25-025-25 HI80-10-010-10 HI68-25-025-50 HI68-25-025-15 HI68-25-015-15 HI68-25-015-50
76429-950EA 3590.22 USD InStock
76429-950 76429-942 76430-070 76429-940 76429-952 76430-072 76429-946 76429-936 76429-944 76429-934 76429-948 76429-938
Avantor® Hichrom HI-DEX DET Beta, GC Columns
Chromatography Columns
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty