Would you like to log in again?
Avantor® Hichrom HI-DEX G-03, GC Columns
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Avantor® Hichrom
HI75-25-015-60 HI75-25-025-15 HI85-10-010-10 HI75-25-015-30 HI75-25-025-60 HI75-25-015-15 HI75-25-015-25 HI75-25-025-30 HI85-10-010-5 HI75-25-025-25 HI75-25-025-50 HI75-25-015-50
76430-036EA 2616.71 USD InStock
76430-036 76430-048 76430-038 76430-090 76430-092 76430-050 76430-040 76430-052 76430-042 76430-044 76430-034 76430-046
Avantor® Hichrom HI-DEX G-03, GC Columns
Chromatography Columns
Unique selectivity for the separation of enantiomers. Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty