Would you like to log in again?
Avantor® Hichrom HI-DEX DMP Beta, GC Columns
Supplier: VWR
Avantor® Hichrom
HI70-25-025-50 HI70-25-015-60 HI82-10-010-10 HI70-25-015-15 HI70-25-015-25 HI70-25-025-25 HI70-25-025-15 HI70-25-015-50 HI70-25-015-30 HI82-10-010-5 HI70-25-025-60 HI70-25-025-30
76429-982EA 3630.81 USD InStock
76429-982 76429-980 76429-992 76429-986 76429-976 76430-080 76429-984 76429-974 76429-988 76429-978 76429-990 76430-078
Avantor® Hichrom HI-DEX DMP Beta, GC Columns
Chromatography Columns
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty