Would you like to log in again?
Avantor® Hichrom HI-DEX DMT Beta, GC Columns
Supplier: VWR
Avantor® Hichrom
HI71-25-015-15 HI83-10-010-5 HI71-25-025-50 HI71-25-015-50 HI83-10-010-10 HI71-25-025-15 HI71-25-025-25 HI71-25-015-60 HI71-25-015-30 HI71-25-025-30 HI71-25-025-60 HI71-25-015-25
76429-994EA 1505.1 USD InStock
76429-994 76430-004 76430-006 76429-998 76430-008 76430-082 76429-996 76430-084 76430-010 76430-000 76430-012 76430-002
Avantor® Hichrom HI-DEX DMT Beta, GC Columns
Chromatography Columns
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty