Very popular nitroterephthalic-acid-modified polyethylene glycol (PEG) column of high polarity for the analysis of free fatty acids, phenols, alcohols, glycols.
- Temperature range*: 35 to 250 °C
- Phase composition: Crossbonded nitroterephthalic-acid-modified polyethylene glycol (PEG)
- Long lifetime
Max temperature limits may change depending on the film thickness.