Would you like to log in again?
Avantor® Hichrom HI-35 HT, GC Columns
Supplier: VWR
Avantor® Hichrom
HI89-53-010-30 HI89-53-010-15 HI89-32-010-30 HI89-32-015-30 HI89-25-010-30 HI89-10-010-10 HI89-32-015-15 HI89-32-010-15 HI89-32-025-30 HI89-32-025-15 HI89-53-025-15 HI89-53-025-30 HI89-25-010-15 HI89-25-015-15 HI89-53-017-30 HI89-53-015-15 HI89-25-015-30 HI89-32-010-10 HI89-10-010-5 HI89-53-015-30 HI89-25-025-15 HI89-53-010-10 HI89-25-025-30
76431-608EA 1010.17 USD InStock
76431-608 76431-606 76431-618 76431-816 76431-604 76431-814 76431-616 76431-594 76431-584 76431-592 76431-582 76431-590 76431-602 76431-614 76431-600 76431-622 76431-612 76431-598 76431-620 76431-610 76431-588 76431-596 76431-586
Avantor® Hichrom HI-35 HT, GC Columns
Chromatography Columns
General purpose column with added high temperature stability for the analysis of Pesticides, PCBs, substituted polar compounds, phenols, etc.
  • Ideal as confrmation column
Max temperature limits may change depending on the film thickness.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty