Would you like to log in again?
Avantor® Hichrom HI-5 HT, GC Columns
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Avantor® Hichrom
HI87-32-015-30 HI87-32-010-10 HI87-25-025-15 HI87-53-025-30 HI87-32-025-15 HI87-25-015-15 HI87-53-017-30 HI87-10-010-10 HI87-53-025-15 HI87-25-025-30 HI87-53-010-15 HI87-25-010-30 HI87-10-010-5 HI87-53-015-15 HI87-53-015-30 HI87-32-025-30 HI87-53-010-10 HI87-25-010-15 HI87-53-010-30 HI873201010G2 HI87-32-010-30 HI87-25-015-30 HI87-32-010-15 HI87-32-015-15
76431-818EA 1194.76 USD InStock
76431-818 76431-628 76431-626 76431-648 76431-638 76431-660 76431-650 76431-186 76431-646 76431-624 76431-636 76431-658 76431-644 76431-820 76431-634 76431-656 76431-664 76431-642 76431-632 76431-654 76431-662 76431-640 76431-652 76431-630
Avantor® Hichrom HI-5 HT, GC Columns
Chromatography Columns
General purpose column with added temperature stability. For the analysis of solvent impurities, PCBs, hydrocarbons, essential oils, semivolatiles, pesticides, phenols and more.
Max temperature limits may change depending on the film thickness.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty