Would you like to log in again?
Avantor® Hichrom HI-8 HT, GC Columns
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Avantor® Hichrom
HI88-10-010-10 HI88-25-015-15 HI883201010G2 HI88-25-010-15 HI88-25-025-15 HI88-10-010-5 HI88-32-015-15 HI88-25-015-30 HI88-32-025-15 HI88-53-010-30 HI88-53-017-30 HI88-53-010-10 HI88-32-010-30 HI88-25-010-30 HI88-53-015-30 HI88-53-025-15 HI88-53-010-15 HI88-32-010-15 HI88-32-015-30 HI88-32-010-10 HI88-25-025-30 HI88-32-025-30 HI88-53-025-30 HI88-53-015-15
76431-708EA 1484.76 USD InStock
76431-708 76431-716 76431-706 76431-824 76431-714 76431-704 76431-682 76431-694 76431-680 76431-692 76431-690 76431-292 76431-822 76431-712 76431-702 76431-710 76431-688 76431-678 76431-700 76431-686 76431-676 76431-698 76431-684 76431-696
Avantor® Hichrom HI-8 HT, GC Columns
Chromatography Columns
High Temperature general purpose column ideal for PCBs compounds, Pesticides, environmental analysis.
Maximum temperature limits may change depending on the film thickness.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty