Really versatile general purpose polar phase for a wide range of applications. Ideal in confirmation analysis. HI-17 MS is an MS grade low-bleeding crossbonded column and assure a long lifetime even under high temperature limits.
- Phase composition: Crossbonded Silphenylene - Methyl Polysiloxane (17MS column selectivity) - MS grade
- Long lifetime
Max temperature limits may change depending on the film thickness.