Proprietary low-polarity and low-bleeding stationary phase; great choice for pesticides, herbicides, polychlorinated biphenyls (PCBs), and polyaromatic hydrocarbons (PAHs). Ideal for GC/MS applications and is also an excellent choice as a confirmation column for semivolatile environmental analysis.
- Phase composition: Crossbonded low-polarity proprietary phase
- Long lifetime
Max temperature limits may change depending on the film thickness.