Really versatile general purpose low-polarity phase for a wide range of applications, including (for example): PCBs, essential oils, pesticides, semivolatiles and many more. HI-5 MS is an MS grade low-bleeding crossbonded column and assure a long lifetime even under high temperature limits.
- Phase composition: crossbonded 5% phenyl - 95% methyl polysiloxane - MS grade
- Long lifetime
Max temperature limits may change depending on the film thickness.