Silphenylene based stationary phase (selectivity equivalent to 5MS columns) to ensure minimum bleeding level at high temperatures. General purpose low-polarity phase for a wide range of applications, including (for example): PCBs, essential oils, pesticides, semivolatiles and many more. HI-5 MS Xil is an MS grade low-bleeding crossbonded column and assure a long lifetime even under high temperature limits.
- Phase composition: Crossbonded Silphenylene - Methyl Polysiloxane (5MS column selectivity) - MS grade
- Long lifetime
Max temperature limits may change depending on the film thickness.