Very popular PEG high polarity phase for a wide range of applications, including (for example): essential oils, flavors and fragrances, alkenols.
- Phase composition: crossbonded polyethylene glycol (PEG)
- Long lifetime
Max temperature limits may change depending on the film thickness.