Would you like to log in again?
Fraser Broth, Modified, Hardy Diagnostics
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
U410
76436-462PK 305.2 USD InStock
76436-462
Fraser Broth, Modified, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Fraser Broth, Modified is recommended for the selective enrichment of Listeria spp. in foods and environmental samples.
  • Heavy duty bottles are tough and sturdy to reduce breakage
  • Easy-to-remove, perforated, shrink wrap ensures product is leak-proof
  • Optically clear bottles are made of clarified polycarbonate for clear viewing
  • Each bottle is easy to read with a clearly marked measuring fill line
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty