Would you like to log in again?
BCYE Agar with VPC (Vancomycin, Polymyxin, Cycloheximide), Hardy Diagnostics
Supplier: Hardy Diagnostics
W79
76436-470PK 49.18 USD InStock
76436-470
BCYE Agar with VPC (Vancomycin, Polymyxin, Cycloheximide), Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Buffered Charcoal Yeast Extract (BCYE) Agar formulations are recommended for use in the cultivation and primary isolation of Legionella spp. in water and other samples suspected of harboring the bacteria.
  • Optically clear polystyrene for distortion free viewing
  • Heavy-Duty to reduce breakage during shipping
  • Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding
  • Packaged in 'breathable' bag that prevents buildup of condensation and excess moisture
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty