Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
3M™ Floor Marking Tapes, Vinyl, 471, Essendant LLC FKA LAGASSE
Supplier: Essendant
3M
MMM471ORG MMM471IWRED MMM471IWYLW
76436-650RL 85.55 USD InStock
76436-650 76436-520 76436-610
3M™ Floor Marking Tapes, Vinyl, 471, Essendant LLC FKA LAGASSE
Tapes
Vinyl Tape 471 is a versatile tape that provides long-lasting floor and safety marking, sharp paint lines on complex jobs, and reliable seals or repairs in a variety of industries.
  • Unique stretch properties enable tape to conform to uneven surfaces without lifting and retracting
  • Rubber adhesive provides excellent holding strength on many surfaces
  • One-piece, clean removal from many surfaces, to reduce clean-up and labor costs
  • Lane and safety marking applications
  • Fine line paint masking
Designed with unique stretch properties, this tape conforms to rough, irregular, curved, and convex surfaces while remaining stretched and without lifting after application. The rubber adhesive offers quick stick and excellent holding strength on many substrates.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty