Would you like to log in again?
WASH STATION PORTABLE 120V 1500WATT
Supplier: LAKESIDE MANUFACTURING INC
Wash Station, Portable, Warm/Cold watersupply with soap, 120v, 1500watt, with adjustable thermostat (110 to 170F), corrosion-resistant bronze tank, Includes built-in 2-1/2-gal electric water heater, SS sink bowl is 10x12x5 deep, Fresh Water: 10gal, Waste Water: 14gal, Size: 32-1/2inx38-1/2x45-1/2in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
WASH STATION PORTABLE 120V 1500WATT 76436-760 9610 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty