Would you like to log in again?
AutoBlock® Fill Filling Station, Environmental Express®
Supplier: Environmental Express
ABF5000 ABF5100
76436-790EA 964.6 USD InStock
76436-790 76436-788
AutoBlock® Fill Filling Station, Environmental Express®
Pumps Drum/Transfer Pumps
Automate the most dangerous step of sample prep!
  • Fit this compact filler in a fume hood for safe and clean operation
  • Utilize Masterflex® pump technology to accurately dispense reagents
  • Intuitive and user-friendly operation through touchscreen or PC
  • Assemble this modular unit within an hour
  • Works with existing HotBlock®, HotBlock® Pro, and HotBlock® 200 digestion systems
Obtain consistent results and reduce preparation time with the AutoBlock Fill. This automated diluter/filler works with any HotBlock digestion system—well formats are already loaded into the software. Use the touchscreen, or connect to a PC, to quickly select which wells need to be filled.
Our AutoBlock Fill Fill Stand is designed for customers who need to prepare samples for multiple HotBlocks. The Fill Stand is constructed of the same, acid-resistant Kydex® material as our HotBlocks. It can hold up to 96 samples at a time, depending on digestion cup size.
Avoid caustic fumes—compact footprint easily fits into a fume hood or benchtop air filtration unit. Masterflex® pump technology accurately dispenses reagents to add confidence to your sample analysis. Eliminate the most dangerous step in metals digestion—acid and reagent addition!
Ordering information: Sample racks are sold separately.
Delivery information: Includes software, integrated pump, PTFE tubing, five 1 L glass reagent bottles, 100 ml waste bottle, and installation hardware.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty