Would you like to log in again?
COD Standards, Environmental Express®
Supplier: Environmental Express
B1030 B1031
76436-808EA 36.56 USD InStock
76436-808 76436-810
COD Standards, Environmental Express®
Standards Environmental Analysis Standards
Controls approved for use in both EPA-compliant and mercury-free vials.
  • Made from primary standard grade potassium hydrogen phthalate
  • Delivered with an SDS and minimum 8 month shelf life (refrigeration required)
  • Easy-to-follow dilution directions printed directly on the label
Certifications: This product complies with the following methods: EPA 410.4, SM 5220D-2011, HACH 8000, ASTM D1252-06, ISO 15705:2002, USGS I-3561-06, BS 6068-2.34:1988, DIN 38414-9, and ISO 6060:1989.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty