Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide concentrate 1.25 N, Snip & Pour™

Supplier: ENVIRONMENTAL EXPRESS
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

C7110
76436-848EA 139.84 USD InStock
76436-848
Sodium hydroxide concentrate 1.25 N, Snip & Pour™
Sodium hydroxide
Deliver precise volume with pre-measured reagents.

  • Easy-to-use, just snip and pour into receiver tube
  • Increase accuracy by eliminating the need to measure and pipette
  • Packaged in convenient PFA tubes


Formula: NaOH
MW: 40 g/mol
MDL Number: MFCD00003548
CAS Number: 1310-73-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty