Would you like to log in again?
FilterMate™ Digestion Cup Filters, Environmental Express®
Supplier: Environmental Express
FilterMate™
SC0404 SC0407 SC0409 SC0408 SC0421
76437-032PK 99.73 USD InStock
76437-032 76437-030 76437-024 76437-028 76437-026
FilterMate™ Digestion Cup Filters, Environmental Express®
Filters Membrane Filter Units
Convenient filtration without a transfer step.
  • Simple to use just press on plunger
  • Filter 15 and 50 ml samples within sample cups
  • No need for separate filtration devices before analyzing samples
Soil, sludge, wastewater, and challenging samples require filtration prior to analysis. FilterMate™ fits directly in the digestion vessel and is pushed through the liquid being filtered to the bottom of the cup with a detachable plunger. After filtration, the plunger is easily removed and discarded while the filter assembly remains in the bottom of the cup. Finished sample can be capped for storage or poured directly into an autosampler vial.
Ordering information: 100 filter assemblies and 100 plungers. Digestion cups sold separately.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty