Would you like to log in again?
FlipMate™ Filtration Assemblies, Environmental Express®
Supplier: Environmental Express
SC0306 SC0301 SC0303 SC0605 SC0302 SC0308 SC0608 SC0602 SC0606 SC0304 SC0651 SC0603 SC0652 SC0604 SC0607 SC0309
76437-042PK 167.35 USD InStock
76437-042 76437-020 76437-054 76437-062 76437-046 76437-058 76437-000 76437-012 76437-056 76437-060 76436-996 76437-050 76437-004 76437-016 76437-038 76437-008
FlipMate™ Filtration Assemblies, Environmental Express®
Filters Membrane Filter Units
The FlipMate™ assemblies easily filter your sample with the use of two threaded digestion cups, a little tubing, and vacuum pressure.
  • Sample is ready in just seconds
  • Convenient, economical method for filtration
  • Produce quality results with clean samples
The sample is placed in one cup, the filter assembly threaded onto the top of the cup, then the cup and filter are flipped, the receiving cup attached, vaccum tubing attached, vaccum pressure applied, and the sample is then pulled from the sample cup, through the filter assembly, and into the empty cup.
Ordering information: Digestions cups, tubing, and vacuum source are not included, filtration assembly only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty