Would you like to log in again?
HotBlock® Digestion Systems, Environmental Express®
Supplier: Environmental Express
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SC100-240 SC196 SC154 SC151-240 SC151 C6002-240 SC154-240 SC196-240 SC150 SC150-240 C6002 SC100
76437-086EA 7118.44 USD InStock
76437-086 76437-064 76437-076 76437-084 76437-074 76437-068 76437-066 76437-078 76437-072 76436-820 76436-822 76437-070
HotBlock® Digestion Systems, Environmental Express®
Digestion Systems Mineralization Workstations
EPA-approved systems that simplify and improve metals prep and digestions.
  • Blocks are PTFE coated for chemical and corrosion resistance
  • Integrated block heater and controller design
  • Set high and low temperature alarm limits
  • Include polycarbonate transfer racks designed to set on the block
The HotBlock® Systems provide faster, cleaner, and more cost-effective metals digestions, and are accepted for EPA Methods. Block heaters have an integrated controller and mat/block design and come in configurations to work directly with the HotBlock digestion cups in 15-, 50-, or 100-mL sizes.
The heating blocks are made from graphite to provide excellent temperature uniformity to each well, and coated in PTFE for chemical and corrosion resistance, and to avoid sample contamination. HotBlock controllers maintain a set point to ±0.2°F (0.1°C) and sample uniformity to ±2.7°F (1.5°C).
Compatible racks are included with each HotBlock and are designed to make sample loading and unloading as easy as possible — racks have the same well design as their compatible HotBlock unit! Racks have a handle molded into each side and are made of clear polycarbonate. The digestion cups are then designed to be compatible with both the HotBlock unit and transfer racks.
Ordering information: Block and transfer rack only. Consumables sold separately.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty