Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SSIbio® Sterile Centrifuge Tubes, Conical and Freestanding, 15 and 50 ml

Supplier: Scientific Specialties

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

2931-S0-500AV 2831-S0-500AV 2830-S0-500AV 2940-S0-500AV 2930-S0-500AV
76437-212CS 277.74 USD InStock
76437-212 76437-210 76437-204 76437-208 76437-206
SSIbio® Sterile Centrifuge Tubes, Conical and Freestanding, 15 and 50 ml
Tubes Centrifuge Tubes
SSIbio Centrifuge Tubes are ideal for everyday centrifugation and storage use.

  • High-clarity, medical-grade polymer
  • Graduations every 1 ml (15 ml tubes) or 5 ml (50 ml tubes) increments
  • Rated to 15000 ×g
  • Temperature range: –80 to 40 °C
  • Sterile and certified free of detectable Dnase, Rnase, DNA, PCR inhibitors, and pyrogen

Tubes include large, white solvent-resistant writing areas and bold, black graduations for easy sample estimation. Flat caps are easy to label.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty