Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Microcentrifuge Tubes for Boiling Applications

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR®
1210-07-201AV 1260-05-201AV 1260-00-201AV 1110-00-101AV 1210-S0-393AV 1210-00-201AV 1310-07-201AV 1260-02-201AV 1310-00-201AV 1210-09-201AV 1110-07-110AV 1260-09-201AV 1110-09-101AV
76332-080CS 1297.1 USD InStock
76332-080 76332-070 76414-040 76437-216 76332-064 76437-214 76332-076 76332-062 76332-074 76414-044 76332-060 76414-042 76332-072
VWR® Microcentrifuge Tubes for Boiling Applications
Tubes Microcentrifuge Tubes
Polypropylene tubes feature superior clarity and a positive seal design that withstands repeated cap closures and will not pop open during boiling.

  • Graduations every 100 µl
  • Tubes fit all standard rotors
  • Certified RNase-, DNase-, and pyrogen-free
  • Certified PCR inhibitor- and human DNA-free
  • Autoclavable
  • 1.5 ml tubes are rated to 30000 ×g when properly supported in the rotor

Temperature range is from –80 to +121 °C (–112 to 250 °F).

Ordering information: Assorted colors include red, blue, green, yellow, and purple tubes.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty