Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Socorex® Qualitix® Sterilization Pouches, DWK Life Sciences
Supplier: DWK Life Sciences
Qualitix®
3210.3040 3210.1326 3210.0816 3210.1936 3210.0926
76437-384CS 161.34 USD InStock
76437-384 76437-382 76437-376 76437-378 76437-380
Socorex® Qualitix® Sterilization Pouches, DWK Life Sciences
Bags Sterilization Pouches
The Qualitix® pouches provide easy and effective wrapping of items requiring sterilization.
  • Robust durable material
  • Large self-sealing strip, easy to close
  • Chevron top seal for easy opening
  • Suitable for steam and EO gas sterilization
  • Two sterilization indicators
  • Five different sizes
Fitting any small to mid-size laboratory device, the Qualitix® single use pouches provide easy and effective wrapping of items requiring sterilization. Made with robust medical grade paper, the pouches tightly close with a self-sealing strip to maintain sterility. Content remains always visible through PET / CPP transparent laminated film.
Certifications: Conformity to ISO 17665-06, ISO 11135, CE.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty