Would you like to log in again?
SafePlus™ MicronRoll Maxi Disposable Wipes, Vileda Professional
Supplier: Vileda Professional
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
166910 166913
76437-394CS 206.81 USD InStock
76437-394 76437-392
SafePlus™ MicronRoll Maxi Disposable Wipes, Vileda Professional
Wipes Disposable Wipes and Towels Wipes for General Applications
The SafePlus™ Mini System is the safe way to dispense single-use microfiber wipes.
  • Effective, quick, and hygienic cleaning of all surfaces, including high-risk areas
  • Reduce the risk of cross-contamination
  • 99.98% of bacteria and germs are removed from surfaces, without disinfectants (certified by 3rd party, BMA Labs, Inc)
  • Sealed polybags fit perfectly into dispensers
  • Easy to use in many areas with different cleaning frequencies, and flexible to store
To achieve a high level of hygiene, the cleaning and disinfection of surfaces is becoming increasingly important. That's why Vileda Professional® offers the most hygienic and safe cleaning solutions for surface cleaning and disinfection.
SafePlus™ wipes achieve the highest level of cleanliness when using a single-use microfiber wipe while cleaning and disinfecting at the same time. The SafePlus™ Maxi dispenser is one of the safest and most innovative wipe dispenser systems available in the market today.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty