Would you like to log in again?
VWR® Surgical Facemasks with Earloops, ASTM 1, 2, and 3
Supplier: VWR
VWR®
SQ3001 SQ1001 SQ2001
76437-414CS 99.88 USD InStock
76437-414 76437-418 76437-416
VWR® Surgical Facemasks with Earloops, ASTM 1, 2, and 3
Masks Surgical Masks
These ASTM rated surgical masks offer various protection levels to fit specific working environments.
  • Multiple layers
  • Comfortable earloops
  • Adjustable nosewire to provide snug fit
  • Inner film layer to maximize breathibility
  • Designed for fluid protection
  • Latex-free
These masks are designed to protect against various ranges of fluid, spray, or debris. When worn properly, they will help to minimze contamination caused by exhaled microorganisms and reduce potential exposure to particulates and fluids.
ASTM Level 1 masks provide a fluid resistance of 80 mm Hg, a BFE of ≥95%, and a PFE of ≥95%.
ASTM Level 2 masks provide a fluid resistance of 120 mm Hg, a BFE of ≥98%, and a PFE of ≥98%.
ASTM Level 3 masks provide a fluid resistance of 126 mm Hg, a BFE of ≥98%, and a PFE of ≥98%.
Certifications: Meets requirements for ASTM F2100-19.
Packaging: Masks are packaged 1000 per case (20 dispensers of 50 masks).
Caution: Prior to use: Check condition of mask, if damaged or soiled, dispose of mask. These masks do not completely eliminate the risk of contracting disease or infection. Adult use only.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty