Would you like to log in again?
High Performance FR Long Sleeve Knit Henley, Hi-Vis Yellow, Lakeland Industries
Supplier: Lakeland Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HSAT29RT-MD HSAT29RT-3X HSAT29RT-XS HSAT29RT-SM HSAT29RT-4XT HSAT29RT-LG HSAT29RT-5XT HSAT29RT-LGT HSAT29RT-XL HSAT29RT-XLT HSAT29RT-4X HSAT29RT-2X HSAT29RT-6X HSAT29RT-2XT HSAT29RT-5X HSAT29RT-MDT HSAT29RT-3XT
76437-630EA 193.27 USD InStock
76437-630 76437-608 76437-606 76437-628 76437-618 76437-622 76437-634 76437-612 76437-620 76437-632 76437-610 76437-626 76437-604 76437-616 76437-624 76437-636 76437-614
High Performance FR Long Sleeve Knit Henley, Hi-Vis Yellow, Lakeland Industries
Workwear
Lightweight, inherently FR Long Sleeve Knit Henley with permanent moisture wicking properties make this the most comfortable FR/AR base layer.
  • Unparalleled CAT 2 protection at 16.4 cal/cm²
  • Lightweight 6.95 oz. inherently FR fabric
  • Excellent breathability
  • Permanent moisture wicking properties
  • Tag free collar
  • Drop shoulder design
  • Left chest pocket
BASE layers are the foundational key to the Lakeland FR/AR Layering System. To be cool during hot summer days, and warm when it's frigid cold outside, wearers need to be dry. BASE layer shirts have the optimal 60/40 blend of hydrophobic and hydrophilic fibers to constantly pull moisture away from the body, move it to the outer surface of the garment, and allow it to dry. Staying dry is the key to keeping wearers comfortable and better protected.
Certifications: NFPA 70E. NFPA 2112 - UL certified. ATPV: 16.4 cal/cm². CAT: 2. ANSI 107 Class 3.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty