Would you like to log in again?
High Performance FR Women's Long Sleeve Knit Henley, Khaki, Lakeland Industries
Supplier: Lakeland Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HSWAT20-XL HSWAT20-XS HSWAT20-3XT HSWAT20-4X HSWAT20-LGT HSWAT20-3X HSWAT20-MD HSWAT20-2XT HSWAT20-XLT HSWAT20-SM HSWAT20-LG HSWAT20-MDT HSWAT20-2X
76437-680EA 174.28 USD InStock
76437-680 76437-670 76437-684 76437-674 76437-682 76437-672 76437-688 76437-666 76437-678 76437-686 76437-664 76437-676 76437-668
High Performance FR Women's Long Sleeve Knit Henley, Khaki, Lakeland Industries
Workwear
Lightweight, inherently FR women's Long Sleeve Knit Henley with permanent moisture wicking properties make this the most comfortable FR/AR base layer.
  • Unparalleled CAT 2 protection at 16.4 cal/cm²
  • Lightweight 6.95 oz. inherently FR fabric
  • Excellent breathability
  • Permanent moisture wicking properties
  • Tag free collar
  • Drop shoulder design
BASE layers are the foundational key to the Lakeland FR/AR Layering System. To be cool during hot summer days, and warm when it's frigid cold outside, wearers need to be dry. BASE layer shirts have the optimal 60/40 blend of hydrophobic and hydrophilic fibers to constantly pull moisture away from the body, move it to the outer surface of the garment, and allow it to dry. Staying dry is the key to keeping wearers comfortable and better protected.
Certifications: NFPA 70E. NFPA 2112 - UL certified. ATPV: 16.4 cal/cm². CAT: 2.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty