Would you like to log in again?
High Performance FR Jackets, Lakeland Industries
Supplier: Lakeland Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
IJKT12ANT06-XLT IJKT12ANT06-MD IJKT12ANT06-XS IJKT12ANT06-LGT IJKT12ANT06-SM IJKT12ANT06-3XT IJKT12ANT06-MDT IJKT12ANT06-XL IJKT12ANT06-3X IJKT12ANT06-5X IJKT12ANT06-4X IJKT12ANT06-2X IJKT12ANT06-4XT IJKT12ANT06-LG IJKT12ANT06-2XT
76437-790EA 511.22 USD InStock
76437-790 76437-794 76437-784 76437-792 76437-806 76437-804 76437-808 76437-798 76437-810 76437-788 76437-796 76437-786 76437-802 76437-800 76437-812
High Performance FR Jackets, Lakeland Industries
Workwear
Layer up and stay warm when donning these High-Performance FR Jackets, featuring inherently FR fabric and permanent moisture wicking properties.
  • Permanent moisture wicking properties
  • 12.3 oz. inherently FR fabric
  • Wind proof and water-resistant
  • Front hand warmer pockets
  • Zipper front closure
Outer layer garments complete the Layering System, providing the comfort and protection wearers demand. Moisture wicking fabrics continue the process of pulling moisture away from the body to the outermost layer so that it can dry, while the wind-resistant and water-resistant fabrics help keep the elements at bay so wearers can focus on the task at hand.
Certifications: NFPA 70E. NFPA 2112 - UL certified. ATPV: 28 cal/cm². CAT: 3.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty