Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 25, 2024. During this time, our site will be not available. We appreciate your patience.
High Performance FR Hi-Vis Jackets, Lakeland Industries
Supplier: Lakeland Industries
IJKT12ANT29RT-SM IJKT12ANT29RT-4XT IJKT12ANT29RT-4X IJKT12ANT29RT-MDT IJKT12ANT29RT-XLT IJKT12ANT29RT-2XT IJKT12ANT29RT-3XT IJKT12ANT29RT-XL IJKT12ANT29RT-2X IJKT12ANT29RT-MD IJKT12ANT29RT-5X IJKT12ANT29RT-XS IJKT12ANT29RT-3X IJKT12ANT29RT-LGT IJKT12ANT29RT-LG
76437-828EA 455.47 USD InStock
76437-828 76437-818 76437-826 76437-816 76437-838 76437-820 76437-842 76437-832 76437-840 76437-830 76437-824 76437-836 76437-814 76437-822 76437-834
High Performance FR Hi-Vis Jackets, Lakeland Industries
Workwear
Layer up and stay warm when donning this High Performance FR Hi-Vis Jacket, featuring inherently FR fabric and permenant moisture wicking properties.
  • Permanent moisture wicking properties
  • 12.3 oz. inherently FR fabric
  • Wind proof and water-resistant
  • Front hand warmer pockets
  • Zipper front closure
Outer layer garments complete the Layering System, providing the comfort and protection wearers demand. Moisture wicking fabrics continue the process of pulling moisture away from the body to the outermost layer so that it can dry, while the wind-resistant and water-resistant fabrics help keep the elements at bay so wearers can focus on the task at hand.
Certifications: NFPA 70E. NFPA 2112 - UL certified. ATPV: 28 cal/cm². CAT: 3. ANSI 107 Class 3.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty