Would you like to log in again?
High Performance FR Men's Dual Hazard Lightweight Pants, Khaki, Lakeland Industries
Supplier: Lakeland Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
IP8WUT20-34-28 IP8WUT20-35-34 IP8WUT20-36-34 IP8WUT20-50-32 IP8WUT20-34-34 IP8WUT20-40-32 IP8WUT20-29-28 IP8WUT20-34-30 IP8WUT20-38-30 IP8WUT20-42-32 IP8WUT20-31-30 IP8WUT20-42-28 IP8WUT20-34-36 IP8WUT20-28-28 IP8WUT20-44-28 IP8WUT20-33-28 IP8WUT20-28-34 IP8WUT20-36-32 IP8WUT20-42-34 IP8WUT20-44-30 IP8WUT20-36-28 IP8WUT20-50-30 IP8WUT20-35-28 IP8WUT20-30-28 IP8WUT20-48-28 IP8WUT20-46-28 IP8WUT20-35-36 IP8WUT20-38-32 IP8WUT20-30-30 IP8WUT20-33-32 IP8WUT20-32-34 IP8WUT20-48-30 IP8WUT20-46-32 IP8WUT20-30-34 IP8WUT20-28-32 IP8WUT20-40-34 IP8WUT20-32-28 IP8WUT20-32-36 IP8WUT20-50-28 IP8WUT20-46-30 IP8WUT20-31-32 IP8WUT20-30-32 IP8WUT20-38-28 IP8WUT20-35-32 IP8WUT20-44-32 IP8WUT20-29-34 IP8WUT20-44-34 IP8WUT20-32-30 IP8WUT20-36-36 IP8WUT20-38-36 IP8WUT20-42-30 IP8WUT20-29-30 IP8WUT20-32-32 IP8WUT20-31-28 IP8WUT20-28-30 IP8WUT20-40-30 IP8WUT20-29-32 IP8WUT20-50-34 IP8WUT20-36-30 IP8WUT20-48-32 IP8WUT20-35-30 IP8WUT20-31-34 IP8WUT20-40-28 IP8WUT20-34-32 IP8WUT20-38-34 IP8WUT20-46-34 IP8WUT20-33-36 IP8WUT20-48-34 IP8WUT20-33-30 IP8WUT20-33-34
76438-036EA 134.49 USD InStock
76438-036 76438-114 76438-038 76438-116 76438-118 76438-084 76438-040 76438-042 76438-086 76438-120 76438-000 76438-088 76438-044 76438-122 76438-046 76438-002 76438-080 76438-082 76438-102 76438-026 76438-104 76438-028 76438-106 76438-108 76438-072 76438-074 76438-030 76438-076 76438-032 76438-110 76438-078 76438-034 76438-112 76438-070 76438-058 76438-014 76438-136 76438-016 76438-138 76438-018 76438-062 76438-020 76438-064 76438-022 76438-066 76438-100 76438-068 76438-024 76438-060 76438-124 76438-004 76438-048 76438-126 76438-006 76438-128 76438-008 76438-094 76438-050 76438-052 76438-096 76438-130 76438-054 76438-098 76438-010 76438-132 76438-012 76438-056 76438-134 76438-090 76438-092
High Performance FR Men's Dual Hazard Lightweight Pants, Khaki, Lakeland Industries
Workwear
High Performance FR Lightweight Pants feature 8 oz. Westex® UltraSoft® fabric that has a unique basketweave blend and provides increased mobility and comfort, making them an excellent choice for arc flash and flash fire protection.
  • Wider waistband for increased comfort
  • Articulated knees offer better mobility
  • Cell phone pocket on right leg
  • Stealth zipper pocket on right leg
  • Crotch gusset for increased comfort
  • Extra material in heal to reduce wear
Lakeland FR Lightweight Pants are made with Westex® UltraSoft® and are NFPA 2112, UL certified, and meet NFPA 70E, CAT 2, making them an excellent choice for arc flash and flash fire protection. The unique basketweave fabric blend provides increased mobility and comfort. Attention to design with features like an articulated knee, wider waistband, and a crotch gusset ensure all day comfort.
Certifications: NFPA 70E. NFPA 2112 - UL certified. ATPV: 9.8 cal/cm². CAT: 2.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty