Would you like to log in again?
High Performance FR Men's Dual Hazard Lightweight Pants, Olive Green, Lakeland Industries
Supplier: Lakeland Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
IP8WUT28-34-34 IP8WUT28-40-30 IP8WUT28-31-34 IP8WUT28-35-32 IP8WUT28-40-36 IP8WUT28-46-32 IP8WUT28-32-32 IP8WUT28-30-28 IP8WUT28-28-30 IP8WUT28-44-30 IP8WUT28-29-34 IP8WUT28-38-34 IP8WUT28-32-28 IP8WUT28-29-30 IP8WUT28-48-28 IP8WUT28-30-32 IP8WUT28-36-36 IP8WUT28-35-34 IP8WUT28-34-36 IP8WUT28-42-32 IP8WUT28-32-34 IP8WUT28-36-28 IP8WUT28-30-30 IP8WUT28-46-30 IP8WUT28-46-28 IP8WUT28-48-32 IP8WUT28-28-34 IP8WUT28-36-32 IP8WUT28-46-34 IP8WUT28-33-34 IP8WUT28-42-34 IP8WUT28-38-28 IP8WUT28-50-28 IP8WUT28-34-28 IP8WUT28-34-32 IP8WUT28-42-30 IP8WUT28-44-34 IP8WUT28-38-30 IP8WUT28-35-28 IP8WUT28-50-34 IP8WUT28-40-34 IP8WUT28-48-30 IP8WUT28-29-28 IP8WUT28-42-28 IP8WUT28-28-32 IP8WUT28-44-28 IP8WUT28-50-30 IP8WUT28-33-30 IP8WUT28-31-32 IP8WUT28-33-32 IP8WUT28-48-34 IP8WUT28-32-30 IP8WUT28-33-28 IP8WUT28-38-32 IP8WUT28-30-34 IP8WUT28-35-30 IP8WUT28-34-30 IP8WUT28-31-30 IP8WUT28-35-36 IP8WUT28-40-32 IP8WUT28-40-28 IP8WUT28-36-30 IP8WUT28-28-28 IP8WUT28-29-32 IP8WUT28-31-28 IP8WUT28-50-32 IP8WUT28-36-34 IP8WUT28-44-32 IP8WUT28-38-36
76438-234EA 144.07 USD InStock
76438-234 76438-158 76438-236 76438-238 76438-160 76438-162 76438-240 76438-164 76438-242 76438-166 76438-244 76438-200 76438-146 76438-224 76438-268 76438-148 76438-226 76438-228 76438-270 76438-194 76438-150 76438-272 76438-196 76438-152 76438-230 76438-274 76438-154 76438-198 76438-276 76438-232 76438-156 76438-190 76438-192 76438-256 76438-212 76438-258 76438-214 76438-216 76438-218 76438-182 76438-260 76438-184 76438-140 76438-262 76438-142 76438-186 76438-220 76438-264 76438-188 76438-144 76438-266 76438-222 76438-180 76438-168 76438-202 76438-246 76438-248 76438-204 76438-206 76438-208 76438-172 76438-250 76438-174 76438-252 76438-176 76438-254 76438-210 76438-178 76438-170
High Performance FR Men's Dual Hazard Lightweight Pants, Olive Green, Lakeland Industries
Workwear
High Performance FR Lightweight Pants feature 8 oz. Westex® UltraSoft® fabric that has a unique basketweave blend and provides increased mobility and comfort, making them an excellent choice for arc flash and flash fire protection.
  • Wider waistband for increased comfort
  • Articulated knees offer better mobility
  • Cell phone pocket on right leg
  • Stealth zipper pocket on right leg
  • Crotch gusset for increased comfort
  • Extra material in heal to reduce wear
Lakeland FR Lightweight Pants are made with Westex® UltraSoft® and are NFPA 2112, UL certified, and meet NFPA 70E, CAT 2, making them an excellent choice for arc flash and flash fire protection. The unique basketweave fabric blend provides increased mobility and comfort. Attention to design with features like an articulated knee, wider waistband, and a crotch gusset ensure all day comfort.
Certifications: NFPA 70E. NFPA 2112 - UL certified. ATPV: 9.8 cal/cm². CAT: 2.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty