Would you like to log in again?
High Performance FR Knit Men's Button Up Shirts, Khaki, Lakeland Industries
Supplier: Lakeland Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ISHAT20-5X ISHAT20-XL ISHAT20-3X ISHAT20-MD ISHAT20-4XT ISHAT20-LG ISHAT20-LGT ISHAT20-2X ISHAT20-4X ISHAT20-XLT ISHAT20-3XT ISHAT20-2XT ISHAT20-MDT ISHAT20-SM
76438-748EA 188.59 USD InStock
76438-748 76438-762 76438-752 76438-764 76438-754 76438-744 76438-766 76438-756 76438-768 76438-746 76438-758 76438-770 76438-760 76438-750
High Performance FR Knit Men's Button Up Shirts, Khaki, Lakeland Industries
Workwear
Featuring Lakeland’s lightweight knit fabric offering permanent moisture-wicking technology. This is the optimal blend for superior performance and protection and comfort.
  • 6.9 oz. lightweight inherently FR fabric
  • Permanent moisture wicking properties
  • Tag-free design
  • Button down collar
  • Athletic cut
  • Two chest pockets
Mid layers are the versatile garments that can help take the chill off for a few hours on a fall morning or provide an extra layer of insulation for those long, frigid winter days. Paired with a Base layer, Mid layers continue to pull moisture away from the body and move it to the outermost garment so it can dry. Lightweight, effective fabrics let Mid layers increase comfort without reducing mobility.
Certifications: NFPA 70E. NFPA 2112 - UL certified. APTV: 16.4 cal/cm². CAT: 2.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty