Would you like to log in again?
Westex® DH 6.5 oz. Women's FR Button Up Shirts, Hi-Vis Yellow, Lakeland Industries
Supplier: Lakeland Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ISHW65DH29RT-2X ISHW65DH29RT-XS ISHW65DH29RT-3X ISHW65DH29RT-2XT ISHW65DH29RT-XL ISHW65DH29RT-SM ISHW65DH29RT-4XT ISHW65DH29RT-4X ISHW65DH29RT-LG ISHW65DH29RT-XLT ISHW65DH29RT-MD ISHW65DH29RT-3XT ISHW65DH29RT-LGT ISHW65DH29RT-MDT ISHW65DH29RT-5X
76438-836EA 224.74 USD InStock
76438-836 76438-826 76438-848 76438-838 76438-828 76438-850 76438-840 76438-830 76438-852 76438-842 76438-854 76438-832 76438-844 76438-834 76438-846
Westex® DH 6.5 oz. Women's FR Button Up Shirts, Hi-Vis Yellow, Lakeland Industries
Workwear
The 6.5 oz. Westex® DH Women's Button Up Shirts have a uniquely woven fabric blend that provides outstanding breathability and advanced moisture management to keep the wearer cooler, drier, and more comfortable.
  • Permanent moisture wicking properties
  • Lightweight 6.5 oz. Westex® DH inherently FR fabric
  • Tag-free collar
  • Athletic cut
  • Button down collar
  • Drop shoulder design
  • Long rounded tail stays tucked in
Mid layers are the versatile garments that can help take the chill off for a few hours on a fall morning or provide an extra layer of insulation for long, frigid winter days. Paired with a Base layer, Mid layers continue to pull moisture away from the body and move it to the outermost garment so it can dry. Lightweight, effective fabrics let Mid layers increase comfort without reducing mobility.
Certifications: NFPA 70E. NFPA 2112 - UL certified. APTV: 8.9 cal/cm². CAT: 2. ANSI 107 Class 3.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty