Would you like to log in again?
High Performance FR Long Sleeve Knit Crew, Khaki, Lakeland Industries
Supplier: Lakeland Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
LSCAT20-4X LSCAT20-6X LSCAT20-MD LSCAT20-3X LSCAT20-2XT LSCAT20-SM LSCAT20-3XT LSCAT20-XLT LSCAT20-LG LSCAT20-XL LSCAT20-XS LSCAT20-MDT LSCAT20-4XT LSCAT20-5X LSCAT20-LGT LSCAT20-2X
76438-946EA 169.9 USD InStock
76438-946 76438-968 76438-958 76438-948 76438-960 76438-950 76438-972 76438-962 76438-952 76438-974 76438-964 76438-954 76438-966 76438-944 76438-956 76438-970
High Performance FR Long Sleeve Knit Crew, Khaki, Lakeland Industries
Workwear
Lightweight, Inherently FR Long Sleeve Knit Crew with permanent moisture wicking properties make this an extremely comfortable FR/AR base layer garment.
  • Unparalleled CAT 2 protection at 16.4 cal/cm²
  • Lightweight 6.95 oz. inherently FR fabric
  • Excellent breathability
  • Permanent moisture wicking properties
  • Tag-free collar
  • Drop shoulder design
  • Left chest pocket
Base layers are the foundational key to the Lakeland FR/AR Layering System. To keep cool during hot summer days, and warm when its frigid cold outside, wearers need to be dry. Base layer shirts have the optimal 60/40 blend of hydrophobic and hydrophilic fibers that constantly pull moisture away from the body, move it to the outer surface of the garment, and allow it to dry. Staying dry is the key to keeping wearers comfortable and better protected.
Certifications: NFPA 70E. NFPA 2112 - UL certified. ATPV: 16.4 cal/cm². CAT: 2.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty