Would you like to log in again?
High Performance FR Waffle Quarter Zip Jackets, Lakeland Industries
Supplier: Lakeland Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
LSCMWZ01-5X LSCMWZ01-XL LSCMWZ01-LG LSCMWZ01-LGT LSCMWZ01-XS LSCMWZ01-XLT LSCMWZ01-SM LSCMWZ01-2XT LSCMWZ01-2X LSCMWZ01-4XT LSCMWZ01-MD LSCMWZ01-MDT LSCMWZ01-3X LSCMWZ01-4X LSCMWZ01-3XT
76439-066EA 180.66 USD InStock
76439-066 76439-044 76439-056 76439-064 76439-042 76439-054 76439-062 76439-040 76439-052 76439-060 76439-050 76439-048 76439-068 76439-046 76439-058
High Performance FR Waffle Quarter Zip Jackets, Lakeland Industries
Workwear
Lakeland brings the incredibly popular waffle cut fabric to High Performance FR with permanent moisture wicking technology and some added stretch. The design ensures wearers will want this hoodie as part of the FR apparel layering system.
  • Pullover quarter zip design
  • Waffle cut textured, inherently FR fabric
  • Permanent moisture wicking properties
  • 8.0 oz. lightweight fabric
Mid layers are the versatile garments that can help take the chill off for a few hours on a fall morning, or provide an extra layer of insulation for long, frigid winter days. Paired with a Base layer, Mid layers continue to pull moisture away from the body and move it to the outermost garment so it can dry. Lightweight, effective fabrics let Mid layers increase comfort without reducing mobility.
Certifications: NFPA 70E. NFPA 2112 - UL certified. APTV: 9.7 cal/cm². CAT: 2.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty