Would you like to log in again?
High Performance FR Sweater-Knit Quarter Zip Jackets, Lakeland Industries
Supplier: Lakeland Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
LSCSK06-XL LSCSK06-2XT LSCSK06-MD LSCSK06-XLT LSCSK06-4X LSCSK06-5X LSCSK06-LG LSCSK06-2X LSCSK06-SM LSCSK06-3X LSCSK06-3XT LSCSK06-MDT LSCSK06-4XT LSCSK06-XS LSCSK06-LGT
76439-080EA 476.52 USD InStock
76439-080 76439-070 76439-092 76439-090 76439-088 76439-078 76439-086 76439-076 76439-098 76439-084 76439-096 76439-074 76439-082 76439-094 76439-072
High Performance FR Sweater-Knit Quarter Zip Jackets, Lakeland Industries
Workwear
Lakeland's new High Performance FR Sweater-Knit Quarter Zip Jackets are a soft and comfortable FR pullover ideal for when the weather turns cold. The jacket features modern, flat seam construction with contrasting black threads with the additional benefits of permanent moisture-wicking technology and incredible horizontal stretch will keep wearers comfortable.
  • Inherently FR sweater knit fabric
  • Permanent moisture wicking technology
  • Quarter zip pullover design
  • Perfect Mid or Outer layer
Outer layer garments complete the Layering System, giving wearers the comfort and protection required. Moisture wicking fabrics continue the process of pulling moisture away from the body to the outermost layer so that it can dry, while the wind-resistant and water-resistant fabrics help keep the elements at bay so wearers can focus on the task at hand.
Certifications: NFPA 70E. NFPA 2112 - UL certified. APTV: 14.0 cal/cm². CAT: 2.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty