Would you like to log in again?
High Performance Women's FR Long Sleeve Knit Crew, Khaki, Lakeland Industries
Supplier: Lakeland Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
LSCWAT20-2XT LSCWAT20-MDT LSCWAT20-XL LSCWAT20-MD LSCWAT20-3X LSCWAT20-XS LSCWAT20-4X LSCWAT20-LGT LSCWAT20-LG LSCWAT20-2X LSCWAT20-SM LSCWAT20-3XT LSCWAT20-XLT
76439-132EA 181.53 USD InStock
76439-132 76439-144 76439-130 76439-142 76439-150 76439-140 76439-138 76439-128 76439-136 76439-126 76439-148 76439-134 76439-146
High Performance Women's FR Long Sleeve Knit Crew, Khaki, Lakeland Industries
Workwear
Lightweight, inherently FR Women's Long Sleeve Knit Crew with permanent moisture wicking properties make this an extremely comfortable FR/AR base layer garment.
  • Unparalleled CAT 2 protection at 16.4 cal/cm²
  • Lightweight 6.95 oz. inherently FR fabric
  • Excellent breathability
  • Permanent moisture wicking properties
  • Tag-free collar
  • Drop shoulder design
Base layers are the foundational key to the Lakeland FR/AR Layering System. To keep cool during hot summer days, and warm when its frigid cold outside, wearers need to be dry. Base layer shirts have the optimal 60/40 blend of hydrophobic and hydrophilic fibers to constantly pull moisture away from the body, move it to the outer surface of the garment, and allow it to dry. Staying dry is the key to keeping wearers comfortable and better protected.
Certifications: NFPA 70E. NFPA 2112 - UL certified. ATPV: 16.4 cal/cm². CAT: 2.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty