Would you like to log in again?
EDX/XEDS Calibration TEM Window Grid, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
76042-01
76439-544PK 724.72 USD InStock
76439-544
EDX/XEDS Calibration TEM Window Grid, Electron Microscopy Sciences
Graticules, Reticles and Grids
Suspended germanium provides a unique calibration standard for x-ray energy dispersive spectroscopy. Since Ge is not typically found in TEM columns, the calibration samples provide a material that cannot be mistaken for instrument components and their signal peaks. The regime in which system peaks normally occur (2 to 9 keV and 11 to 20 keV) is devoid of peaks from the Ge.
  • 20 nm thick germanium (Ge) coating on microporous 20 nm thick, low-stress silicon nitride (SiN)
  • 2 µm pores on 1:1 pitch grid pattern
  • 100 µm thick frame, fits 3 mm sample holders
  • (1) 500×500 µm window
The Ge is suspended across 2 µm pores that are patterned on a grid of 20 nm thick silicon nitride.
The single 500×500 µm window is compatible with high tilt angle tomography, since at 70° of tilt, the thin and beveled 100 µm silicon frame allows you to use a ~50×50 µm region within the center of the window from any rotational orientation.
These EDX calibration standards were developed in partnership with Dr. Nestor J. Zaluzec from the Electron Microscopy Center and the Center for Nanoscale Materials at Argonne National Laboratory.
Applications in detector energy axis and energy resolution calibration, detector window transmission evaluation, detector solid angle measurements, electron optical instrument system peak measurements, and specimen.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty