Would you like to log in again?
QUANTIFOIL® R Holey Carbon Films on London Finder
Supplier: Electron Microscopy Sciences
LFH7100AR1-2NM LFH6100AR1-2NM LFH7100AR35-2 LFH6100AR1.3 LFH6100CR1 LFH15100CR1-2 LFH15100CR35-2 LFH6100CR4-2NM LFH7100CR2 LFH7100AR1.3-2NM LFH6100AR35-2 LFH2100CR4-2NM LFH15100CR2-2 LFH6100AR2-2NM LFH2100AR4 LFH2100AR4-2NM LFH15100AR1.3 LFH6100AR2 LFH2100AR1 LFH7100AR2 LFH2100AR35-2 LFH6100CR35-2 LFH7100CR1.3-2 LFH6100CR35 LFH2100AR1-2NM LFH15100AR2 LFH7100AR1.3 LFH15100CR1 LFH15100AR1-2 LFH15100CR1.3 LFH2100CR35-2 LFH2100CR1-2NM LFH7100AR4 LFH7100AR4-2NM LFH2100CR35 LFH15100CR35 LFH2100CR1.3 LFH2100AR1.3 LFH7100CR4-2NM LFH15100AR4-2 LFH2100CR1 LFH15100CR1.32 LFH6100CR4 LFH2100AR2-2NM LFH2100CR2-2NM LFH7100CR4 LFH7100CR35-2 LFH6100CR1-2NM LFH6100CR1.3 LFH7100AR2-2NM LFH15100AR4 LFH6100AR35 LFH2100AR1.3-2 LFH2100AR2 LFH7100AR1 LFH15100AR1 LFH6100AR1 LFH15100CR2 LFH15100CR4 LFH6100AR1.3-2 LFH7100CR1-2NM LFH2100CR2 LFH7100CR35 LFH6100CR1.3-2 LFH2100CR4 LFH15100AR1.32 LFH7100CR1 LFH2100CR1.3-2 LFH15100AR35-2 LFH7100CR2-2NM LFH6100AR4 LFH7100AR35 LFH2100AR35 LFH6100CR2-2NM LFH15100AR35 LFH15100AR2-2 LFH6100CR2 LFH15100CR4-2 LFH6100AR4-2NM LFH7100CR1.3
76440-092PK 1922.56 USD InStock
76440-092 76440-094 76440-050 76440-090 76439-954 76439-998 76439-952 76439-996 76440-016 76439-994 76439-950 76439-992 76440-018 76440-012 76440-056 76440-058 76440-014 76440-052 76440-096 76439-958 76440-098 76440-010 76439-956 76440-054 76440-080 76440-082 76439-990 76440-008 76439-988 76439-986 76440-004 76440-048 76439-984 76440-006 76439-982 76440-000 76440-044 76440-088 76440-046 76440-002 76440-040 76439-948 76440-084 76439-946 76440-086 76440-042 76440-070 76440-072 76439-980 76439-976 76439-974 76439-972 76440-038 76439-970 76440-034 76440-078 76440-036 76440-074 76440-030 76440-032 76439-978 76440-076 76440-060 76439-966 76439-964 76439-962 76440-026 76440-104 76440-028 76439-960 76440-066 76440-022 76440-100 76440-024 76440-068 76440-102 76440-062 76440-064 76439-968 76440-020
QUANTIFOIL® R Holey Carbon Films on London Finder
Graticules, Reticles and Grids
QUANTIFOIL® is a carbon film for electron microscopy or low-energy electron point source microscopy.
QUANTIFOIL® is a perforated support foil with pre-defined hole size, shape and arrangement that has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) when compared with conventional holey film. QUANTIFOIL® with circular holes is used in cryo-electron tomographic reconstruction. The roundness of the holes is advantageous with respect to the formation of an ice layer of constant thickness.
The hole size chosen depends on the magnification used, and on whether or not one wishes to include support film in the image. Assessment of the image quality is easier when foil is included in the picture, because the power spectrum of a foil is stronger than that of unsupported ice.
The surface properties of QUANTIFOIL holey carbon support film, especially the wetting properties, may have to be adapted according to one's particular requirements. Untreated aging QUANTIFOIL tends to be hydrophobic. Hydrophillicity of the foil can be achieved by glow discharging in residual air or by metal coating.
R 1.2/1.3 has 1.2 µm circular holes with a space of 1.3 µm between the holes.
R 2/1: Hole size is 2 µm. Space between holes is 1 µm. Center to center is 3 µm.
R 2/2: Hole size is 2 µm. Space between holes is 2 µm. Center to center is 4 µm.
2/4: Hole size is 2 µm. Space between holes is 4 µm. Center to center is 6 µm.
3.5/1: Hole size is 3.5 µm. Space between holes is 1 µm. Center to center is 4.5 µm.
Ordering information: QUANTIFOIL® is delivered as a carbon foil; it can be reinforced with plastic film. The grids are packed with their coated side toward the center of the grid box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty