Would you like to log in again?
QUANTIFOIL® R 1 Holey Carbon Films on Grids
Supplier: Electron Microscopy Sciences
Q410AR-12 Q210CR-12 Q210CR-14 Q225NR-12 Q420NR-14 Q410NR-12 Q450CR-12 Q410AR-14 Q250AR-12 Q325AR-12 Q210NR-12 Q310AR-14 Q250NR-12 Q310NR-14 Q2100CR-12 Q225CR-12 Q220AR-14 Q410CR-12 Q310NR-12 Q410NR-14 Q420AR-14 Q210AR-14 Q210NR-14 Q250CR-12 Q220CR-14 Q310CR-12 Q420CR-14 Q410CR-14 Q210AR-12 Q325NR-12 Q225AR-12 Q350NR-12 Q310AR-12 Q320CR-14 Q325CR-12 Q425CR-14 Q450NR-12 Q350AR-12 Q425NR-12 Q450AR-12 Q350CR-12 Q320NR-14 Q425AR-12 Q320AR-14 Q220NR-14 Q425CR-12 Q310CR-14
76441-116PK 598.92 USD InStock
76441-116 76441-214 76441-278 76440-908 76441-030 76441-310 76441-034 76440-628 76441-132 76440-976 76440-778 76440-676 76440-158 102773-334 76440-678 76440-810 76440-876 76440-710 76440-430 76440-154 76440-398 76440-310 76440-212 76440-574 76440-294 76440-570 76441-148 76441-124 76441-084 76441-182 76441-244 76440-718 76440-624 76440-328 76440-208 76440-726 76441-082 76440-500 76440-302 76440-744 76440-468 76440-842 76440-980 76440-366 76440-260 103306-342 76440-262
QUANTIFOIL® R 1 Holey Carbon Films on Grids
Graticules, Reticles and Grids
QUANTIFOIL® is a carbon film for electron microscopy or low-energy electron point source microscopy.
  • 1 µm circular holes and a space between the holes of 2 µm
  • Mounted on copper, nickel or gold grids
  • Foil is 12 nm thick
  • Thicknesses between 10 and 25 nm upon request
QUANTIFOIL® is a perforated support foil with pre-defined hole size, shape and arrangement that has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) when compared with conventional holey film. QUANTIFOIL® with circular holes is used in cryo-electron tomographic reconstruction. The roundness of the holes is advantageous with respect to the formation of an ice layer of constant thickness.
The hole size chosen depends on the magnification used, and on whether or not one wishes to include support film in the image. Assessment of the image quality is easier when foil is included in the picture, because the power spectrum of a foil is stronger than that of unsupported ice.
QUANTIFOIL® R 1/2 has 1 µm circular holes and a space of 2 µm between the holes. The QUANTIFOIL® types with small holes (e.g. 1.2 µm) were designed for magnifications around and higher 50000x.
QUANTIFOIL® R 1/4 has a hole size of 1 µm. Space between holes is 4 µm. The R 1/4 may be preferred over R 1.2/1.3, when an increased tolerance with respect to the position of beam, and a larger beam diameter are desired, such as in the case of automated image acquisition. holes (e.g. 1.2 µm) were designed for magnifications around and higher 50000x.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty