Would you like to log in again?
Quantifoil® SiO₂ Holey Carbon Films on Grids
Supplier: Electron Microscopy Sciences
Q225AS3510 Q350AS3510 Q210AS355 Q450AS3510 Q325AS355 Q425CS355 Q250AS355 Q425NS3510 Q410AS3510 Q450NS355 Q450AS355 Q325CS355 Q325NS355 Q250NS3510 Q3100CS355 Q310CS355 Q4100AS355 Q310CS3510 Q4100NS3510 Q350CS3510 Q410AS355 Q250AS3510 Q3100AS355 Q4100CS355 Q210NS355 Q225CS3510 Q425AS355 Q310AS3510 Q310NS355 Q250NS355 Q350CS355 Q410NS355 Q350AS355 Q250CS3510 Q350NS355 Q425NS355 Q4100AS3510 Q410NS3510 Q225-NS7 Q310AS7 Q425CS3510 Q225NS355 Q325CS3510 Q325AS3510 Q310NS7 Q225CS355 Q310CS7 Q210CS355 Q325NS3510 Q250CS355 Q410CS7 Q410NS7 Q450CS3510 Q3100AS3510 Q4100NS355 Q210CS7 Q310NS3510 Q2100NS3510 Q225AS355 Q4100CS3510 Q210NS7 Q350NS3510 Q2100CS3510 Q2100NS355 Q450CS355 Q310AS355 Q2100CS355 Q3100NS3510 Q225NS3510 Q450NS3510 Q210CS3510 Q3100NS355 Q210AS7 Q410AS7 Q2100AS355 Q210AS3510 Q425AS3510 Q3100CS3510 Q210NS3510 Q2100AS3510 Q410CS355 Q410CS3510
76440-290PK 189.59 USD InStock
76440-290 76440-490 76441-172 76440-926 76440-606 76441-012 76441-010 76440-658 76440-536 76440-416 76440-418 76440-456 76440-136 76441-170 76440-458 76440-656 76440-898 76440-534 76440-132 76440-134 76440-492 76440-130 76441-204 76441-328 76441-206 76441-008 76441-326 76440-518 76440-958 76440-768 76440-602 76440-800 76440-924 76440-802 76440-604 76440-766 76440-242 76440-286 76440-288 76440-520 76440-244 76440-240 76440-120 76440-190 76440-390 76440-706 76441-112 76441-232 76440-826 76441-110 76441-230 76440-828 76440-118 76440-954 76440-956 76440-356 76440-952 76440-550 76440-354 76440-552 76441-302 76441-268 76441-300 76441-108 100491-236 76441-064 76441-062 76441-266 76441-066 76440-548 76440-900 76440-702 76440-866 76440-704 76440-940 76440-942 76440-546 76440-864 76440-188 76440-660 76440-388 76440-186
Quantifoil® SiO₂ Holey Carbon Films on Grids
Graticules, Reticles and Grids
QUANTIFOIL® is a carbon film for electron microscopy or low-energy electron point source microscopy.
QUANTIFOIL® with square holes and relatively narrow bars can be used in electron microscopy to support a thin carbon film, which by itself is too fragile to span a grid square. Alternatively, this holey film can directly support an object larger than the holes.
QUANTIFOIL® S 7/2 has a hole size of 7×7 µm. Space between holes is 2 µm. Center to center is 9 µm.
QUANTIFOIL S 35/5 hole size is 35 µm. Space between holes is 5 µm. Center to center is 40 µm.
QUANTIFOIL S 35/10 hole size is 35 µm. Space between holes is 10 µm. Center to center is 45 µm.
Ordering information: QUANTIFOIL® is delivered as a carbon foil; it can be reinforced with plastic film. The grids are packed with their coated side toward the center of the grid box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty