Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

QUANTIFOIL® R 10 Holey Carbon Films on Grids

Supplier: Electron Microscopy Sciences
Q325NR1020 Q350CR1020 Q250AR1020 Q225CR105 Q310CR1010 Q410AR1020 Q225AR1020 Q250CR1020 Q210CR1010 Q410CR105 Q310NR1010 Q350NR1010 Q450CR1010 Q425AR105 Q425CR105 Q425AR1020 Q350NR105 Q350CR1010 Q4100AR1020 Q450CR105 Q225AR1010 Q225NR1020 Q325AR1010 Q250CR1010 Q450NR1020 Q3100NR1010 Q225CR1020 Q210NR105 Q450NR1010 Q410AR1010 Q250AR1010 Q325AR105 Q210NR1020 Q350AR1010 Q425CR1020 Q250NR1020 Q250AR105 Q350NR1020 Q425NR105 Q410NR105 Q410CR1010 Q450AR1010 Q3100AR105 Q425CR1010 Q210AR1020 Q425AR1010 Q210CR105 Q310CR1020 Q250CR105 Q250NR105 Q310NR1020 Q410NR1020 Q225CR1010 Q350CR105 Q325NR105 Q225AR105 Q325CR105 Q210NR1010 Q310AR1020 Q325CR1020 Q225NR105 Q310AR105 Q310CR105 Q450CR1020 Q410AR105 Q450AR105 Q325AR1020 Q325CR1010 Q210CR1020 Q410NR1010 Q410CR1020 Q425NR1020 Q310AR1010 Q425NR1010 Q325NR1010 Q210AR1010 Q350AR105 Q310NR105 Q450AR1020 Q250NR1010 Q225NR1010 Q210AR105 Q450NR105 Q350AR1020
76441-212PK 541.52 USD InStock
76441-212 76440-808 76441-178 76441-210 76440-804 76441-176 76440-806 76440-972 76440-258 76440-774 76440-776 76440-974 76440-496 76440-254 76440-498 76440-970 76440-256 76440-772 103306-572 76440-494 76441-208 103307-050 103307-012 76441-240 76440-838 76441-242 76440-206 76440-202 76440-566 76440-324 76440-840 76441-080 76440-568 76440-204 76440-326 76440-364 76440-564 76440-322 76440-360 76440-362 76441-238 76441-272 76440-904 76441-276 76441-078 76440-906 76441-274 76441-076 76440-836 76440-874 76440-870 76440-672 76440-396 76440-872 76440-674 76441-308 76440-150 76440-392 76440-670 76440-394 76440-152 76441-028 76441-026 76441-146 76441-306 76441-304 76440-618 76441-024 76441-144 76440-738 76441-142 76440-426 76440-902 76440-428 76440-742 76441-180 76440-620 76440-466 76440-148 76440-424 76440-622 76440-462 76440-740 76440-464
QUANTIFOIL® R 10 Holey Carbon Films on Grids
Graticules Reticles and Grids
QUANTIFOIL® is a carbon film for electron microscopy or low-energy electron point source microscopy.

  • Mounted on copper, nickel or gold grids
  • Foil is 12 nm thick
  • Thicknesses between 10 and 25 nm upon request

QUANTIFOIL® is a perforated support foil with pre-defined hole size, shape and arrangement that has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) when compared with conventional holey film. QUANTIFOIL® with circular holes is used in cryo-electron tomographic reconstruction. The roundness of the holes is advantageous with respect to the formation of an ice layer of constant thickness.

The hole size chosen depends on the magnification used, and on whether or not one wishes to include support film in the image. Assessment of the image quality is easier when foil is included in the picture, because the power spectrum of a foil is stronger than that of unsupported ice.

QUANTIFOIL® R 10/5 has a hole size of 10 µm. Space between holes is 5 µm. Center to center is 15 µm.
QUANTIFOIL® R 10/10 has a hole size of 10 µm. Space between holes is 10 µm. Center to center is 20 µm.
QUANTIFOIL® R 10/20 has a hole size of 10 µm. Space between holes is 20 µm. Center to center is 30 µm.

Ordering information: QUANTIFOIL® is delivered as a carbon foil; it can be reinforced with plastic film. The grids are packed with their coated side toward the center of the grid box.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty