Would you like to log in again?
Buffered Saline Prefilled Tubes, Azer Scientific
Supplier: Azer Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ES45048
76441-352CS 1046.75 USD InStock
76441-352
Buffered Saline Prefilled Tubes, Azer Scientific
Buffers Electrophoresis Buffers Washing Buffers
This sterile 0.85% buffered saline solutions are liquid mediums recommended for use in qualitative procedures that require the use of an isotonic diluent.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty