Would you like to log in again?
NoTrax™ 355 Shoe Sanitizing and Drying Mat Systems, Justrite®
Supplier: Justrite
NoTrax®
355SC048BL 355S002700 355SC018CH 355SC024CH 355S002500 355SC024BL
76441-368EA 475.35 USD InStock
76441-368 76441-378 76441-372 76441-370 76441-376 76441-374
NoTrax™ 355 Shoe Sanitizing and Drying Mat Systems, Justrite®
Floor Mats Permanent Floor Mats
The shoe sanitizer mat helps to disinfect shoes and provides an absorbent mat to quickly dry shoes.
  • Effective shoe cleaner prior to sanitizing shoes
  • A simple, safe solution to help disinfect shoes from spreading contaminants
  • Easy instruction to sanitize shoes
  • Vinyl backed to prevent slipping
  • Adhesive backed replaceable mats for hassle-free cleaning
  • Easy to set up and move
It's never been easier to disinfect shoes with this 2 or 3 zone shoe sanitizer systems. This disinfectant shoe mat not only will safely clean and sanitize shoes, but quickly dry them the moment you step out.
Cleaning Zone 1 (only avaliable in the 3 Zone Systems) acts as the shoe scraper to help remove dirt, grime and organic matter to better sanitize shoes.
Disinfecting Zone 2 has a removable tray to be filled with a disinfect mat solution. While saturating the bottom of shoe soles, the mat insert has fibers help guide the solution into deep shoe treds.
Drying Zone 3 uses a highly absorbent berber to provide quick drying and slip prevention after disinfecting shoes.
The shoe sanitizer system comes with instructional messaging, making it ideal for entrances for schools, businesses, facilities, and even homes. The system contains adhesive backed disposable carpets for quick and easy cleaning and installation. Disinfecting shoes provides the peace of mind by reducing the amount of bacteria and virus spread from the soles of shoes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty