Would you like to log in again?
Intelsius® Pathopouch, DGP Intelsius, LLC
Supplier: DGP Intelsius
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BP004 BP006 BP002 BP001 BP005 BP003 BP008 BP007
76441-388BX 18.46 USD InStock
76441-388 76441-390 76441-394 76441-382 76441-392 76441-386 76441-396 76441-384
Intelsius® Pathopouch, DGP Intelsius, LLC
Bags Sample Bags
The Intelsius® Pathopouch range are 95 kPa flexible pressure vessels designed to meet with transport regulations.
  • 95 kPa plastic secondary packaging
  • Outer pouch for paperwork
  • Leak-proof
They are leak-proof and suitable to hold a wide range of primary containers, such as blood vials, specimen containers, swabs, blood collection bags and medical devices.
Certifications: ISTA Certified Class 6.2 Category A & B
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty