Would you like to log in again?
Corning® Ultra-Low Attachment 25 cm² Rectangular Canted Neck Cell Culture Flask with Vent Cap, Corning
Supplier: Corning
Corning®
4616
76441-540CS 612.26 USD InStock
76441-540
Corning® Ultra-Low Attachment 25 cm² Rectangular Canted Neck Cell Culture Flask with Vent Cap, Corning
Media for Microbiology Prepared Media for Microbiology
Features a covalently bound hydrogel layer that effectively inhibits cellular attachment and minimizes protein absorption, enzyme activation, and cellular activation.
  • Surface is non-cytotoxic, biologically inert, and nondegradable
  • Lot numbers for ease in traceability
  • 100% integrity tested
  • Sterilized by gamma irradiation
  • Nonpyrogenic
The Corning vessels feature a biologically inert ultra-low attachment surface that minimizes cell attachment and promotes the formation of 3D multicellular spheroids employed in cancer research, stem cell biology, and drug screening.
Minimizes cell attachment, protein absorption, and enzyme activation.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty