Would you like to log in again?
Excellence XPR Analytical Balances, METTLER TOLEDO®
Supplier: Mettler-Toledo
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
30355342 30355411 30594488 30469150 30469154 30355419 30594483 30594483 30594488
76292-090EA 19260 USD InStock
76292-090 CA76443-138 76292-152 76443-138 CA76443-136 76292-088 76292-110 76292-092 76443-136
Excellence XPR Analytical Balances, METTLER TOLEDO®
Balances and Scales
The next generation of Analytical weighing has arrived. The XPR analytical balances provide ultimate accuracy and quality assurance functions that are second to none.
  • Results protocol
  • Storage of methods
  • Balance ready StatusLight
  • LevelControl: Level warning with graphical levelling guide
  • Ionizer module ready, LabX ready and calibry ready
  • Upgradable to automatic powder and / or liquid dosing
The XPR Analytical Balances, succesor to the XPE Analytical Balances, delivers ultimate weighing performance and a range of intelligent quality assurance functions that fulfill the most stringent requirements. You can depend on XPR to deliver right-first-time results. The patented StaticDetect™ technology, used in combination with the optional ionizing module, offers a complete electrostatic detection and elimination system to ensure accurate weighing results and highest process security.
Features with 7" color touchscreen (glove compatible), ergoClip for direct dosing, SmartSens for touch-less door operation, SmartTrac dosing guide automatic draft shield doors, proFACT advanced internal adjustment, MinWeigh warning function and SmartGrid weighing pan.
It has seamless process, tolerance profiles, efficient operations, adjustment and routine test history, change history function, user management and Active Temperature Control system (ATC™).
Ordering information: Optional modules and accessories make it easy to customize your XPR analytical balance to perfectly fit your process needs.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty